Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 3.8mL, Max Recovery™, Screw Top, Amber Autosampler Vials and 100 EA matching Screw Caps with a Silicone Rubber / PTFE Liner in the Black solid Caps. 1/PK.
Catalog No. 502271088
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-1088 Supplier Microsolv Technology Corporation Supplier No. 9534SSPA13
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.