Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa, fitted in the Robo Style, Blue Caps. Vials and Caps are Pre-assembled. 1/PK.
Catalog No. 502270065
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
This item is not returnable. View return policy
Catalog No. 50-227-0065 Supplier Microsolv Technology Corporation Supplier No. 9502SE1CPAX
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.