Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 300ul, Fused Insert with a wide tip, Screw Top, Clear RSA Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with White Silicone Rubber / Red PTFE Soft-Guard Septa, bonded in the Blue Caps. 1 PK.
Catalog No. 502271092
Encompass_Preferred
Easy Purchase Pack™ Vial and Cap Kits are a convenient way to order and keep inventory of autosampler vials with their matching caps organized. The kit packaging will open and re-close making it useful for keeping vial and cap types together and clean during use and storage.
This item is not returnable. View return policy
Catalog No. 50-227-1092 Supplier Microsolv Technology Corporation Supplier No. 9532S1ECPWTRS
Easy Purchase Pack™ Vial and Cap Kits are a convenient way to order and keep inventory of autosampler vials with their matching caps organized. The kit packaging will open and re-close making it useful for keeping vial and cap types together and clean during use and storage.