Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 2ml, Push Top, Amber Autosampler Vials and 100 EA matching, translucent, polyethylene Push Caps for Micro-Insert. 1/PK.
Catalog No. 502271263
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. Shell Vials made with 1st Hydrolytic, Pharmaceutical grade Borosilicate glass with outer dimensions 31.5x11.6mm and High purity polyethylene Push Top Caps.
This item is not returnable. View return policy
Catalog No. 50-227-1263 Supplier Microsolv Technology Corporation Supplier No. 9502CASVEPP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. Shell Vials made with 1st Hydrolytic, Pharmaceutical grade Borosilicate glass with outer dimensions 31.5x11.6mm and High purity polyethylene Push Top Caps.