Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Amber Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Red PTFE / White Silicone Rubber / Red PTFE Septa, bonded in the Blue Caps, 1/PK.
Catalog No. 502270617
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0617 Supplier Microsolv Technology Corporation Supplier No. 9532S2AP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.