Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear Autosampler Vials with Write-On Patch and 100 EA matching Screw Caps with Clear Silicone Rubber / Clear PTFE, Pre-Slit, ultra-pure Septa, fitted in the Light Blue Caps. 1PK
Catalog No. 502270327
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0327 Supplier Microsolv Technology Corporation Supplier No. 9509S3TWCPAQ
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.