Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 2ml, Snap Top, Amber RSA™ Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Snap Caps with Silicone Rubber / PTFE Pre-Slit, ultra-pure Septa, fitted in the Light Blue Caps. 1/PK.
Catalog No. 502270340
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0340 Supplier Microsolv Technology Corporation Supplier No. 9509C3WAPRS
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.