Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 3.8ml, Maximum Recovery, Screw Top, Clear Deactivated Library Storage Vials and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa bonded in the Black Caps. 1/PK.
Catalog No. 502271089
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
This item is not returnable. View return policy
Catalog No. 50-227-1089 Supplier Microsolv Technology Corporation Supplier No. 9534S1DCP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.