Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 1.8ml, Maximum Recovery, Snap Top, Clear Autosampler Vials and 100 EA matching Snap Caps with ultra pure, PTFE, fitted in the Blue Caps. 1/PK.
Catalog No. 502270606
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0606 Supplier Microsolv Technology Corporation Supplier No. 9512C5CP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.