Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 40mL, Screw Top, Glass, EPA Style vials, Amber and 100 EA matching Screw Caps with Silicone Rubber / White PTFE Septa, fitted in the white caps. 1/PK.
Catalog No. 502271389
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-1389 Supplier Microsolv Technology Corporation Supplier No. 9704005WP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.