Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 40ml, Center Draining, Screw Top, Clear Autosampler Vials and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa fitted in the White Caps. Vials and Caps are Pre-assembled. 1/PK.
Catalog No. 502271388
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
This item is not returnable. View return policy
Catalog No. 50-227-1388 Supplier Microsolv Technology Corporation Supplier No. 970401CP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.