Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 144 EA 4ml, Screw Top, Clear Storage Vials and 100 EA matching, Attached Screw Caps with a Rubber / PTFE Liner fitted in the Black, Knurled, Phenolic Caps. Easy Purchase Pack. 144/CS.
Catalog No. 502271392
Encompass_Preferred
These 4ml Vials are made with 1st Hydrolytic, TYPE 33 Borosilicate Glass with 13-425mm threads that a supplied in egg crate style corrugated tray, boxes. Closed, or solid, Phenolic caps are supplied and can tolerate higher temperatures than polypropylene and are harder. Caps and Vials are supplied assembled.
This item is not returnable. View return policy
Catalog No. 50-227-1392 Supplier Microsolv Technology Corporation Supplier No. 9700401
These 4ml Vials are made with 1st Hydrolytic, TYPE 33 Borosilicate Glass with 13-425mm threads that a supplied in egg crate style corrugated tray, boxes. Closed, or solid, Phenolic caps are supplied and can tolerate higher temperatures than polypropylene and are harder. Caps and Vials are supplied assembled.