Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Amber RSA Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure, Pre-Slit Septa, fitted in the Light Blue Caps. 1/PK.
Catalog No. 502270114
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0114 Supplier Microsolv Technology Corporation Supplier No. 9532S3APRS
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.