Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 300ul, Fused Insert, Screw Top, Clear Deactivated Autosampler Vials with Write on Patch and fill lines and 100 EA matching Screw Caps with Clear Silicone Rubber / Clear Tefzel, ultra-pure Septa, fitted in the Blue Caps. 1/PK.
Catalog No. 502270119
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0119 Supplier Microsolv Technology Corporation Supplier No. 9532S060DCP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.