Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 300ul, Screw Top, Clear polypropylene LCMS Compatible Autosampler Vials and 100 EA matching Screw Caps with Silicone Rubber / PTFE, ultra-pure Septa, fitted in the Light Blue Caps. 1/PK.
Catalog No. 502270127
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0127 Supplier Microsolv Technology Corporation Supplier No. 9532SMS10P
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.