Learn More
Microsolv Technology Corporation Vial & Cap Kit. 100 EA 2ml, Screw Top, Hydrophobic, Clear Autosampler Vials and matching Solid Black Caps with Clear Silicone Rubber / PTFE Liner. 1 PK.
Catalog No. 502270158
Encompass_Preferred
These RSA Glass Autosampler Vial & Cap Kit Packs (Easy Purchase Packs) are supplied with Clear RSA Vials that have been treated with RSA-Pro X™ Technology to prevent their already pristine surface from adsorbing stubbornly sticky compounds that have affinity for the glass structure. The Hydrophobic surface is not only non-adsorbing for most compounds but is extremely stable even in the presence of Water. The hydrolytic stability will last for years compared to Silanized Vials that will usually degrade in the presence of Water. This product is manufactured in an ISO 9001:2015 Certified facility. The supplied, Solid Top matching Black, AQR™ Screw Caps have the most inert Silicone Rubber / PTFE Liner available; for total Vial and Cap inertness. Often chosen for storage or shipping of standards, proteins, peptides, enzymes, natural products and other precious compounds. These treated vials are only sold in RSA-Pro X ™ Kits.
This item is not returnable. View return policy
Catalog No. 50-227-0158 Supplier Microsolv Technology Corporation Supplier No. 9509SKDCPPX
These RSA Glass Autosampler Vial & Cap Kit Packs (Easy Purchase Packs) are supplied with Clear RSA Vials that have been treated with RSA-Pro X™ Technology to prevent their already pristine surface from adsorbing stubbornly sticky compounds that have affinity for the glass structure. The Hydrophobic surface is not only non-adsorbing for most compounds but is extremely stable even in the presence of Water. The hydrolytic stability will last for years compared to Silanized Vials that will usually degrade in the presence of Water. This product is manufactured in an ISO 9001:2015 Certified facility. The supplied, Solid Top matching Black, AQR™ Screw Caps have the most inert Silicone Rubber / PTFE Liner available; for total Vial and Cap inertness. Often chosen for storage or shipping of standards, proteins, peptides, enzymes, natural products and other precious compounds. These treated vials are only sold in RSA-Pro X ™ Kits.