Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 2ml, Snap Top, Amber Autosampler Vials with Write-On Patch and 100 EA matching Snap Caps with Silicone Rubber / PTFE Pre-Slit, Septa fitted in the Blue Caps. Vials and Caps are Pre-assembled. 1/PK.
Catalog No. 502269924
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
Catalog No. 50-226-9924 Supplier Microsolv Technology Corporation Supplier No. 9502C3WAPA
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.