Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 2ml, Crimp Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Crimp Caps with Red Rubber / PTFE Septa, fitted in the Silver Caps. 1/PK
Catalog No. 502269928
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-226-9928 Supplier Microsolv Technology Corporation Supplier No. 9502C40WAP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.