Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 2ml, Screw Top, Clear RSA™ Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Silicone Rubber / PTFE Pre-Slit Septa, bonded in the Blue Caps, 1/PK.
Catalog No. 502270083
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0083 Supplier Microsolv Technology Corporation Supplier No. 9509S3XWCPRS
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.