Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 300ul, Fused Insert, Snap Top, Clear Autosampler Vials with Write on Patch and fill lines and 100 EA matching Snap Caps with Red Rubber / PTFE Septa fitted in the Red Caps. 1/PK.
Catalog No. 502270120
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0120 Supplier Microsolv Technology Corporation Supplier No. 9532C4CRP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.