Learn More
Microsolv Technology Corporation Vial & Cap Kit includes 100 EA 300ul, Fused Insert, Snap Top, Clear RSA™ Autosampler Vials with Write on Patch and fill lines and 100 EA matching Snap Caps with Clear AQR Silicone Rubber / Clear PTFE, ultra-pure Septa, fitted in the Light Blue Caps. 1/PK.
Catalog No. 502270122
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.
This item is not returnable. View return policy
Catalog No. 50-227-0122 Supplier Microsolv Technology Corporation Supplier No. 9532C1CPRS
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps. The kit packaging will open and re-close making it useful for keeping vial and cap types matched, organized and clean during use.