Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 100 EA 4ml, Screw Top, Amber Autosampler Vials with Write-On Patch and fill lines and 100 EA matching Screw Caps with Silicone Rubber / PTFE Septa, bonded in the Black Caps, 1/PK.
Catalog No. 502270132
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
This item is not returnable. View return policy
Catalog No. 50-227-0132 Supplier Microsolv Technology Corporation Supplier No. 9504S1WAP
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.