Learn More
Microsolv Technology Corporation Vial & Cap Kit Includes 1000 EA 300ul, Screw Top, Clear polypropylene Autosampler Vials and 1000 EA matching Screw Caps with Silicone Rubber / PTFE Septa, bonded in the Blue Caps, 1/PK.
Supplier:  Microsolv Technology Corporation 9532S0P1CK
Encompass_Preferred
Easy Purchase Packs are a convenient way to order and inventory autosampler vials with their matching caps.
Catalog No. 50-227-0144