Learn More
CEM Corporation VIAL CAPS 35ML 100/PK
SDP
Catalog No. NC0759153
Encompass
35mL/80mL SP-D Plus Pressure Caps. Includes quantity 100 of 35mL/80mL vial caps with TFM septa. Compatible with Discover SP-D.
Catalog No. NC0759153 Supplier CEM Corporation Supplier No. 909355
35mL/80mL SP-D Plus Pressure Caps. Includes quantity 100 of 35mL/80mL vial caps with TFM septa. Compatible with Discover SP-D.