Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Vial Cleaning Brush

Catalog No. 09054122 Shop All Tintometer Group Products
Change view
Click to view available options
Length (Metric):
110 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
09-054-122 110 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-054-122 Supplier Lovibond™ Supplier No. 380230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Vial Cleaning Brush is designed to clean the inside of a Lovibond™ sample cell without scratching or damaging glass.

  • Designed to clean the inside of a sample cell.

Specifications

Type Vial Cleaning Brush
Length (English) 4.3 in.
Length (Metric) 110 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.