Learn More
Savillex Corporation Vial Cleaning System
SDP
Catalog No. NC1666872
Encompass
For cleaning PFA vials and labware by heated soaking. Maximum surface temperature of hotplate for vial cleaning system is 450°F (230°C). System components: cleaning container, cleaning container lid, sealing ring, vent nut assembly, pour spout assembly, ½” cap nut and silicone grip mat. Included silicone grip mat holds container in place when sealing ring is tightened or removed.
Catalog No. NC1666872 Supplier Savillex Corporation Supplier No. 55001
For cleaning PFA vials and labware by heated soaking. Maximum surface temperature of hotplate for vial cleaning system is 450°F (230°C). System components: cleaning container, cleaning container lid, sealing ring, vent nut assembly, pour spout assembly, ½” cap nut and silicone grip mat. Included silicone grip mat holds container in place when sealing ring is tightened or removed.