Learn More
Perkin Elmer US LLC 2 mL Autosampler Convenience Kit - 9 mm Screw Top Clear Polypropylene Vial with PTFE/Silicone Pre-Slit Septa, 100/pk
Catalog No. NC1778593
Encompass_Preferred
Vial Top Type Vial Volume Capacity Vial Material Neck I.D. Size Cap Type Septa Material Septa Type Quantity Screw Top 2 mL Clear Polypropylene 9 mm Blue Cap PTFE/Silicone Pre-Slit 100
Catalog No. NC1778593 Supplier Perkin Elmer US LLC Supplier No. N9301736
Vial Top Type Vial Volume Capacity Vial Material Neck I.D. Size Cap Type Septa Material Septa Type Quantity Screw Top 2 mL Clear Polypropylene 9 mm Blue Cap PTFE/Silicone Pre-Slit 100