Learn More
Electron Microscopy Sciences V-Vials Borosilicate Clear Vial with Cap 0.3 ml
SDP
Catalog No. 501899981
Encompass_Preferred
These vials are made from Wheaton-33 low extractable borosilicate glass and they safeguard against any change of product pH caused by alkali, which can be extracted from ordinary glass. Wheaton V-Vials are exceptionally sturdy vials with a conical interior bottom. Their special features insure maximum security and economy in handling, storing and shipping of costly micro and semi-micro samples.
V-Vials are ideal for Micro Mixing, Homogenization, Centrifugation, Concentration, Extraction, Derivatization, Radiosynthesis, Hybridization, Anaerobic Reactions, and Catalytic Hydrogenation.
All components can be autoclaved. They come packed 12 vials in a hinged lid storage box. Graduated; clear; screw cap has solid top and TFE-faced white (styrene butadiene) rubber liner.
Volume: 0.3 ml
Catalog No. 50-189-9981 Supplier Electron Microscopy Sciences Supplier No. 7264701
These vials are made from Wheaton-33 low extractable borosilicate glass and they safeguard against any change of product pH caused by alkali, which can be extracted from ordinary glass. Wheaton V-Vials are exceptionally sturdy vials with a conical interior bottom. Their special features insure maximum security and economy in handling, storing and shipping of costly micro and semi-micro samples.
V-Vials are ideal for Micro Mixing, Homogenization, Centrifugation, Concentration, Extraction, Derivatization, Radiosynthesis, Hybridization, Anaerobic Reactions, and Catalytic Hydrogenation.
All components can be autoclaved. They come packed 12 vials in a hinged lid storage box. Graduated; clear; screw cap has solid top and TFE-faced white (styrene butadiene) rubber liner.
Volume: 0.3 ml