Learn More
Chemglass Life Sciences Vial, Collection, 4 Dram, 25 x 52mm, Glass Only
SDP
Supplier:  Chemglass Life Sciences CG186950
Encompass_Preferred
Vial, Collection, 4 Dram, 25 x 52mm, Glass Only. Part of the CG-1869-01 Bill-Board set. The Bill-Board set has been designed to simplify and expedite multiple, solid-phase syntheses. Up to 6 reactions can be conducted simultaneously in fritted glass reaction vessels. The Bill-Board equipment keeps the solid-phase reactions organized in a grid (for potential combinatorial reaction sequences). Also, the system simplifies the essential steps of solid-phase synthesis which involve repeated cycles of reactions and washings, followed by a final cleavage, product collection and solvent evaporation. The Bill-Boards can also be used for any procedures that require multiple filtrations (i.e. simultaneous silica gel chromatography).
Catalog No. 50-153-1270