Learn More
Chemglass Life Sciences 10mL Conical Reaction Vial, Thin-Wall, Minum-Ware
SDP
Catalog No. 50873530
Encompass_Preferred
10mL Conical Reaction Vial, Thin-Wall, Minum-Ware. Vials are thin walled allowing more efficient heat transfer. Conical bottoms are designed for optimum sample retrieval. With 14/10 threaded outer joint. Supplied complete with open-top screw cap (MW-102-33), PTFE faced septa (MW-104-33) and Viton O-ring (MW-100-08).
Catalog No. 50-873-530 Supplier Chemglass Life Sciences Supplier No. MW5403
10mL Conical Reaction Vial, Thin-Wall, Minum-Ware. Vials are thin walled allowing more efficient heat transfer. Conical bottoms are designed for optimum sample retrieval. With 14/10 threaded outer joint. Supplied complete with open-top screw cap (MW-102-33), PTFE faced septa (MW-104-33) and Viton O-ring (MW-100-08).