Learn More
Chemglass Life Sciences 5mL Conical Reaction Vial, Thin-Wall, Minum-Ware
SDP
Catalog No. 501532688
Encompass_Preferred
5mL Conical Reaction Vial, Thin-Wall, Minum-Ware. Vials are thin walled allowing more efficient heat transfer. Conical bottoms are designed for optimum sample retrieval. With 14/10 threaded outer joint. Supplied complete with open-top screw cap (MW-102-33), PTFE faced septa (MW-104-33) and Viton O-ring (MW-100-08).
Catalog No. 50-153-2688 Supplier Chemglass Life Sciences Supplier No. MW5402
5mL Conical Reaction Vial, Thin-Wall, Minum-Ware. Vials are thin walled allowing more efficient heat transfer. Conical bottoms are designed for optimum sample retrieval. With 14/10 threaded outer joint. Supplied complete with open-top screw cap (MW-102-33), PTFE faced septa (MW-104-33) and Viton O-ring (MW-100-08).