Learn More
Trajan Scientific L-MARK Brand, Vial, Crimp, Amber, 2mL, 12x32mm mm, Certified Mass Spec Quality, 11 mm, Case of 1000
Catalog No. 501251021
Encompass_Preferred
L-MARK Brand, Vial, Crimp, Amber, 2mL, 12x32mm mm, Certified Mass Spec Quality, 11 mm, Case of 1000. Made in the USA.
Catalog No. 50-125-1021 Supplier Trajan Scientific Supplier No. LPP.10002CMS10
L-MARK Brand, Vial, Crimp, Amber, 2mL, 12x32mm mm, Certified Mass Spec Quality, 11 mm, Case of 1000. Made in the USA.