Learn More
Cole-Parmer Nalgene 5000-1020 Cryogenic vial; 1.5 mL, 25/pack
Supplier:  Cole-Parmer 0675494
Encompass_Preferred
Nalgene 5000-1020 Cryogenic vial; 1.5 mL, 25/packExternal threads and long skirt length reduce risk of contaminationPolypropylene vials with HDPE closures are self-standingVials withstand temperatures from –196 to 120°CWhite marking area aids in sample identification. Graduated on the side. Order colored cap inserts separately.Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64 (order separately).
Catalog No. NC0298924