Learn More
Chemglass Life Sciences Vial, Cryo, 2.0mL, Sterile, Self-Standing, External Thread, Silicone Washer Seal
SDP
Catalog No. NC1064535
Encompass_Preferred
Vial, Cryo, 2.0mL, Sterile, Self-Standing, External Thread, Silicone Washer Seal, Pack of 100
Catalog No. NC1064535 Supplier Chemglass Life Sciences Supplier No. CLS476202A
Vial, Cryo, 2.0mL, Sterile, Self-Standing, External Thread, Silicone Washer Seal, Pack of 100