Learn More
Thomas Scientific Sample Tubes, Ext. threads, 2 mL, Self-standing, w/o cap
SDP
Catalog No. 501461680
Encompass_Preferred
Made of polypropylene Designed for the storage and transportation of biological material. Manufactured from non-toxic polypropylene, the tube provides strength and clarity and exhibits some unique design features. The vial has external threads, providing a smooth and uniform inner surface, thus reducing the risk of contamination. Tubes can be autoclaved (121 °C) in upright position with caps loosened. Height of tube is with cap. Round bottom tubes only can be centrifuged up to 17,000g. Order caps separately.
This item is not returnable. View return policy
Catalog No. 50-146-1680 Supplier Thomas Scientific Supplier No. 1219G15
Made of polypropylene Designed for the storage and transportation of biological material. Manufactured from non-toxic polypropylene, the tube provides strength and clarity and exhibits some unique design features. The vial has external threads, providing a smooth and uniform inner surface, thus reducing the risk of contamination. Tubes can be autoclaved (121 °C) in upright position with caps loosened. Height of tube is with cap. Round bottom tubes only can be centrifuged up to 17,000g. Order caps separately.