Learn More
MilliporeSigma™ Supelco™ Vial Decapper, Pliers-Type
Catalog No. 111011242 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
11 mm seals
13 mm seals
20 mm seals
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-101-1242 11 mm seals
11-101-1243 13 mm seals
11-101-1244 20 mm seals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-1242 Supplier MilliporeSigma™ Supelco™ Supplier No. 33281
Only null left
Add to Cart
Add to Cart
Vial Decapper, pliers-type is used for quick and easy removal of a aluminum seal.
TRUSTED_SUSTAINABILITY
Specifications
Description Decapper, pliers style
No. per Pack 1/Pk.
For Use With (Equipment) 11 mm seals