Learn More
Labcon North America 25mL Sample Dilution Vials with Attached Caps, in Sectioned Box
Catalog No. 50101611
Encompass_Preferred
Vial, Dilution; 25mL; Labcon; For Beckman Coulter style blood cell counters and benchtop dilutions; Particle-free; ATP, Endotoxin free (non-pyrogenic); With attached leak-proof natural polyethylene cap; Clear virgin polystyrene; Used in blood cell counter; 144 units/sectioned box, 5 boxes/case; Made in USA
Catalog No. 50-101-611 Supplier Labcon North America Supplier No. 20305500009
Vial, Dilution; 25mL; Labcon; For Beckman Coulter style blood cell counters and benchtop dilutions; Particle-free; ATP, Endotoxin free (non-pyrogenic); With attached leak-proof natural polyethylene cap; Clear virgin polystyrene; Used in blood cell counter; 144 units/sectioned box, 5 boxes/case; Made in USA