Learn More
TWD Scientific LLC Vial, Cryogenic, Twd Biox, 2.0Ml, External Thread, Clear O-Ring Screw Cap, Sterile, Polypropylene, Self-Standing Conical Bottom, Writing Patch, Marked Graduations, 2000 Vials/Case.
SDP
Catalog No. 502285046 Shop All TWD Scientific LLC Products
Encompass_Preferred
2.0Ml Clear Polypropylene Sterile Self-Standing Conical Bottom Cryogenic Vials With External Thread And Clear O-Ring Screw Caps Assembled. Vials Have Writing Patch And Marked Graduations. 50 Vials/Bag, 10 Bags/Pack, 4 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5046 Supplier TWD Scientific LLC Supplier No. BX818204
2.0Ml Clear Polypropylene Sterile Self-Standing Conical Bottom Cryogenic Vials With External Thread And Clear O-Ring Screw Caps Assembled. Vials Have Writing Patch And Marked Graduations. 50 Vials/Bag, 10 Bags/Pack, 4 Packs/Case, Price/Case