Learn More
Bellco Glass, Inc. Vial F/B,1.5mL
SDP
Catalog No. 501945623
Encompass_Preferred
DescriptionThis vial is made of clear borosilicate glass and has flat bottom. Sample capacity of 1.5mL, 9mm in diameter, and 44mm in height. Vials compatible with our Multi-Tier Plate System. 100 vials per case.Multi-Tier Plate System, is an adaptable, and reusable system, which features a versatile 96 well base plate that can secure vials ranging from 0.5mL to 2.5mL and the ability to isolate individual vials as needed.The ANSI footprint ensures compatibility with all autosamplers and robotic systems.Product DetailsColor: Clear GlassSize: 9x44mmVolume: 1.5mLSilanized: NoBottom: FlatCase Quantity: 100Country Of Origin: USA
Catalog No. 50-194-5623 Supplier Bellco Glass, Inc. Supplier No. 451001030
DescriptionThis vial is made of clear borosilicate glass and has flat bottom. Sample capacity of 1.5mL, 9mm in diameter, and 44mm in height. Vials compatible with our Multi-Tier Plate System. 100 vials per case.Multi-Tier Plate System, is an adaptable, and reusable system, which features a versatile 96 well base plate that can secure vials ranging from 0.5mL to 2.5mL and the ability to isolate individual vials as needed.The ANSI footprint ensures compatibility with all autosamplers and robotic systems.Product DetailsColor: Clear GlassSize: 9x44mmVolume: 1.5mLSilanized: NoBottom: FlatCase Quantity: 100Country Of Origin: USA