Learn More
Trajan Scientific L-MARK Brand Vial, Headspace, Crimp, 10-mL, Clear, round bottom, 100 Pack
Catalog No. 501037369
Encompass_Preferred
L-MARK Brand Vial, Headspace, Crimp, 10-mL, Clear, round bottom, beveled top, 20mm, 22.6x46 100 Pack. Made in the USA.
Catalog No. 50-103-7369 Supplier Trajan Scientific Supplier No. LPP.10751
L-MARK Brand Vial, Headspace, Crimp, 10-mL, Clear, round bottom, beveled top, 20mm, 22.6x46 100 Pack. Made in the USA.