Learn More
Trajan Scientific L-MARK Brand Vial, 10.0 mL, Amber, Headspace, 20mm Crimp, 22.6x46, 100 Pack
Supplier:  Trajan Scientific LPP.10765
Encompass_Preferred
L-MARK Brand Vial 10.0 mL Vial, Amber, Headspace, 20mm Crimp, Bevelled Top, Flat Bottom, 22.6x46100 Pack. Made in the USA.
Catalog No. 50-109-0160