Learn More
Trajan Scientific L-MARK Brand, Clear Vial, Headspace, Crimp, 20mL, Clear, Standard finish, Round bottom, 20mm, 22.6x75.5, 100 Pack
Catalog No. 501090147
Encompass_Preferred
L-MARK Brand 20.0 mL Clear Vial, 22.6 x 75.5, 20mm Crimp Standard Top/Long Neck, Radius Bottom. Made in the USA.
Catalog No. 50-109-0147 Supplier Trajan Scientific Supplier No. LPP.10732
L-MARK Brand 20.0 mL Clear Vial, 22.6 x 75.5, 20mm Crimp Standard Top/Long Neck, Radius Bottom. Made in the USA.