Learn More
Trajan Scientific La-Pha-Pack , Vial, Headspace, Crimp, 20mL, Clear, Radius Bottom, 23x75.5, 100 Pack. Made in Germany.
Catalog No. 501037356
Encompass_Preferred
La-Pha-Pack , Vial, Headspace, Crimp, 20mL, Clear, Radius Bottom, 23x75.5, 100 Pack. Made in Germany.
Catalog No. 50-103-7356 Supplier Trajan Scientific Supplier No. LAP.20090297
La-Pha-Pack , Vial, Headspace, Crimp, 20mL, Clear, Radius Bottom, 23x75.5, 100 Pack. Made in Germany.