Learn More
Trajan Scientific La-Pha-Pack , Vial, Headspace, Clear, Crimp, 5mL, 20x38mm, Flat bottom, 100 Pack. Made in Germany.
Catalog No. 501037360
Encompass_Preferred
La-Pha-Pack , Vial, Headspace, Clear, Crimp, 5mL, 20x38mm, Flat bottom, 100 Pack. Made in Germany.
Catalog No. 50-103-7360 Supplier Trajan Scientific Supplier No. LAP.20090801
La-Pha-Pack , Vial, Headspace, Clear, Crimp, 5mL, 20x38mm, Flat bottom, 100 Pack. Made in Germany.