Learn More
Trajan Scientific L-MARK Brand Headspace Vial 10mL 18mm Screw Thread 22.6x46 Round Bottom, 100 Pack
Catalog No. 501090168
Encompass_Preferred
L-MARK Brand Vial, 10.0 mL, Amber,Headspace, 18mm Screw Thread, 18mm S/T, Radius Bottom (CTC), 22.6x46100 Pack. Made in the USA.
Catalog No. 50-109-0168 Supplier Trajan Scientific Supplier No. LPP.10786
L-MARK Brand Vial, 10.0 mL, Amber,Headspace, 18mm Screw Thread, 18mm S/T, Radius Bottom (CTC), 22.6x46100 Pack. Made in the USA.