Learn More
Chemglass Life Sciences Vial, High Recovery, 3.7mL, Clear, GPI 13-425, 15 x 45mm, Complete with Open-Top Cap And Septa
SDP
Catalog No. 50143892
Encompass_Preferred
High recovery vial with a conical bottom for complete sample recovery. These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Available complete with open-top caps or vials only.
Catalog No. 50-143-892 Supplier Chemglass Life Sciences Supplier No. CG491630
High recovery vial with a conical bottom for complete sample recovery. These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Available complete with open-top caps or vials only.