Learn More
Chemglass Life Sciences Vial Complete, Sample, 7.3mL, Clear, High Recovery, 17x60mm, GPI 15-425, with Open-Top Cap and Septa
SDP
Catalog No. 50143889
Encompass_Preferred
Vial Complete, Sample, 7.3mL, Clear, High Recovery, 17x60mm, GPI 15-425, with Open-Top Cap and Septa. High recovery vial with a conical bottom for complete sample recovery. These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Complete with open-top caps.
Catalog No. 50-143-889 Supplier Chemglass Life Sciences Supplier No. CG491631
Vial Complete, Sample, 7.3mL, Clear, High Recovery, 17x60mm, GPI 15-425, with Open-Top Cap and Septa. High recovery vial with a conical bottom for complete sample recovery. These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Complete with open-top caps.