Learn More
Chemglass Life Sciences Vial Only, Sample, 11.4mL, Clear, High Recovery, 19x70mm, GPI 15-425
SDP
Catalog No. 50230670
Encompass_Preferred
Vial Only, Sample, 11.4mL, Clear, High Recovery, 19x70mm, GPI 15-425. High recovery vial with a conical bottom for complete sample recovery. These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Vial only.
Catalog No. 50-230-670 Supplier Chemglass Life Sciences Supplier No. CG491603
Vial Only, Sample, 11.4mL, Clear, High Recovery, 19x70mm, GPI 15-425. High recovery vial with a conical bottom for complete sample recovery. These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Vial only.