Learn More
Agilent Technologies VIAL, HIGH RECOVERY, SNAP TOP,
Catalog No. NC2145643
Encompass
Vial, high recovery, snap top, 1.5 mL with 30 µL reservoir, clear, 100/pk. Vial size: 12 x 32 mm (11 mm cap)
This item is not returnable. View return policy
Catalog No. NC2145643 Supplier Agilent Technologies Supplier No. 51834510
Vial, high recovery, snap top, 1.5 mL with 30 µL reservoir, clear, 100/pk. Vial size: 12 x 32 mm (11 mm cap)