Learn More
Analytical Sales & Services Vial Kit: 11mL (3 Dram) 19 x 65mm Amber Screw Vials with Solid Teflon Lined Screw Caps, 144/Pack
Catalog No. 5000806085
Encompass
Advantage™ Assembled Vial Kit. Contains: 11mL (3 Dram) 19 x 65mm Amber Screw Vials with Solid Teflon Lined Screw Caps. Pack of 144 vials.
Catalog No. 50-008-06085 Supplier Analytical Sales & Services Supplier No. ADV69308A
Advantage™ Assembled Vial Kit. Contains: 11mL (3 Dram) 19 x 65mm Amber Screw Vials with Solid Teflon Lined Screw Caps. Pack of 144 vials.